Arsenic (V) oxide ≥99.9% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
14668-14
14668-22
CAAA14668-14EA
244.9
CAD
CAAA14668-14
CAAA14668-22
Arsenic (V) oxide ≥99.9% (metals basis)
Arsenic (V) oxide
MDL: MFCD00003434
Freely soluble in water and alcohol
Purity: 99.9%
CAS No.: 1303-28-2
Molecular Formula: As2O5
Formula Weight: 229.79
Physical Form: -100 Mesh Powder
Density: 4.32
Appearance: White
Odor: Odorless
Melting Point: 315° dec.
Solubility: Freely soluble in water and alcohol
Merck Reference: 14,798
UN No.: 1559
MDL No.: MFCD00003434
Formula:
As₂O₅ Melting Pt: 315 °C (dec.) Density: 4.32 Storage Temperature: Ambient |
MDL Number:
MFCD00003434 CAS Number: 1303-28-2 EINECS: 215-116-9 UN: 1559 ADR: 6.1,II Merck Index: 14,798 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...