Tin 99+% (metals basis), foil, Length: 150 mm, Width: 270 mm, Thickness: 0.33 mm (0.013 in)

Supplier: Thermo Scientific Chemicals

14636-RD 14636-RE
CAAA14636-RDEA 284.4 CAD
CAAA14636-RD CAAA14636-RE
Tin 99+% (metals basis), foil, Length: 150 mm, Width: 270 mm, Thickness: 0.33 mm (0.013 in)
Tin

MDL: MFCD00133862


Purity: 99+% CAS No.: 7440-31-5 Physical Form: ≈100g/15x27cm Odor: Odorless MDL No.: MFCD00133862

Formula: Sn
Boiling Pt: 2270 °C (1013 hPa)
Melting Pt: 232 °C
Density: 7.31 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133862
CAS Number: 7440-31-5
EINECS: 231-141-8
Merck Index: 12,09587

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR