Titanium(IV) oxide rutile ≥99.99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
14631-09
14631-18
14631-30
CAAA14631-09EA
170.64
CAD
CAAA14631-09
CAAA14631-18
CAAA14631-30
Titanium(IV) oxide rutile ≥99.99% (metals basis)
Titanium (IV) oxide rutile
MDL: MFCD00011269
Purity: 99.99%
CAS No.: 1317-80-2
Molecular Formula: TiO2
Formula Weight: 79.90
Physical Form: Powder
Density: 4.23
Appearance: White
Odor: Odorless
Melting Point: 1830-1850°
Boiling Point: 2500-3000°
Merck Reference: 14,9472
MDL No.: MFCD00011269
Formula:
O₂Ti Boiling Pt: 2500…3000 °C Melting Pt: 1830…1850 °C Density: 4.23 Storage Temperature: Ambient |
MDL Number:
MFCD00011269 CAS Number: 1317-80-2 EINECS: 215-282-2 Merck Index: 14,9472 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...