Zinc ≥99% (metals basis), mossy ≤ 2.5 cm (0.98 in)

Supplier: Thermo Scientific Chemicals

14629-30 14629-A1
CAAA14629-30EA 51.67 CAD
CAAA14629-30 CAAA14629-A1
Zinc ≥99% (metals basis), mossy ≤ 2.5 cm (0.98 in)
Zinc

MDL: MFCD00011291


Purity: 99% CAS No.: 7440-66-6 Odor: Odorless MDL No.: MFCD00011291

Formula: Zn
Boiling Pt: 908 °C (1013 hPa)
Melting Pt: 420 °C
Density: 7.14 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011291
CAS Number: 7440-66-6
EINECS: 231-175-3
Merck Index: 12,10255

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR