Zirconium ≥99.2% (metals basis excluding Hf), wire (max. 4.5% Hf), Ø 1.14 mm (0.045 in)

Supplier: Thermo Scientific Chemicals

14627-G2 14627-G6 14627-H2
CAAA14627-G2EA 81.37 CAD
CAAA14627-G2 CAAA14627-G6 CAAA14627-H2
Zirconium ≥99.2% (metals basis excluding Hf), wire (max. 4.5% Hf), Ø 1.14 mm (0.045 in)
Zirconium

MDL: MFCD00011303


Purity: 99.2% CAS No.: 7440-67-7 Physical Form: ≈6.64g/m Odor: Odorless MDL No.: MFCD00011303

Formula: Zr
MW: 91.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011303
CAS Number: 7440-67-7
EINECS: 231-176-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR