Indium antimonide ≥99.999% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

14626-04 14626-09
CAAA14626-04EA 78.21 CAD
CAAA14626-04 CAAA14626-09
Indium antimonide ≥99.999% (metals basis)
Indium antimonide

MDL: MFCD00016146


Purity: 99.999% CAS No.: 1312-41-0 Molecular Formula: InSb Formula Weight: 236.57 Physical Form: 6mm & Down Pieces Density: 5.75 at m.p. Melting Point: 535° Merck Reference: 14,4948 UN No.: 1549 MDL No.: MFCD00016146

Formula: InSb
MW: 236.58
Melting Pt: 527
Density: 5.75 (m.p.
Storage Temperature: Ambient
MDL Number: MFCD00016146
CAS Number: 1312-41-0
EINECS: 215-192-3
UN: 1549
ADR: 6.1 III
Merck Index: 14,4948

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR