Neodymium tribromide hydrate ≥99.9% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals
Warning

14606-14 14606-22
CAAA14606-14EA 68.1 CAD
CAAA14606-14 CAAA14606-22
Neodymium tribromide hydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Neodymium tribromide hydrate

MDL: MFCD00748137


Purity: 99.9% CAS No.: 29843-90-1 Molecular Formula: NdBr3.xH2O Formula Weight: 383.97(anhy) Physical Form: Crystalline

Formula: Br₃Nd·H₂O
Storage Temperature: Ambient
MDL Number: MFCD00748137
CAS Number: 29843-90-1
EINECS: 236-897-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR