Samarium tribromide hexahydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Samarium tribromide hexahydrate, Samarium(III) bromide hexahydrate
14603-14
14603-22
CAAA14603-14EA
94.96
CAD
CAAA14603-14
CAAA14603-22
Samarium tribromide hexahydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Samarium tribromide hexahydrate
MDL: MFCD00192430
Purity: 99.9%
CAS No.: 13517-12-9
Molecular Formula: SmBr3.6H2O
Formula Weight: 498.15 (390.06anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
MDL No.: MFCD00192430
Formula:
Br₃Sm·6H₂O MW: 498.16 g/mol Melting Pt: >640 °C Storage Temperature: Ambient |
MDL Number:
MFCD00192430 CAS Number: 13517-12-9 EINECS: 237-347-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...