Terbium (III) sulfate octahydrate ≥99.9% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Diterbium trisulphate octahydrate, Diterbium trisulfate octahydrate

14596-04 14596-09 14596-18
CAAA14596-04EA 80.11 CAD
CAAA14596-04 CAAA14596-09 CAAA14596-18
Terbium (III) sulfate octahydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Terbium (III) sulphate octahydrate

MDL: MFCD00149872


Purity: 99.9% CAS No.: 13842-67-6 Molecular Formula: Tb2(SO4)3.8H2O Formula Weight: 750.19 (606.05anhy) Physical Form: Crystalline Melting Point: 360° -8H2O MDL No.: MFCD00149872

Formula: O₁₄S₃Tb₂·8H₂O
MW: 782.16 g/mol
Melting Pt: 360 °C (-8H₂O)
Storage Temperature: Ambient
MDL Number: MFCD00149872
CAS Number: 13842-67-6
EINECS: 237-213-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR