Holmium trinitrate pentahydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Scientific Chemicals
|
Danger
|
14588-09
14588-18
CAAA14588-09EA
170.64
CAD
CAAA14588-09
CAAA14588-18
Holmium trinitrate pentahydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Holmium trinitrate pentahydrate
MDL: MFCD00149737
Purity: 99.9%
CAS No.: 14483-18-2
Molecular Formula: Ho(NO3)3.5H2O
Formula Weight: 441.02 (350.94anhy)
Physical Form: Crystalline Aggregates
Sensitivity: Hygroscopic
UN No.: 1477
MDL No.: MFCD00149737
Formula:
HoN₃O₉·5H₂O Melting Pt: 83.74…84.62 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149737 CAS Number: 14483-18-2 EINECS: 233-438-8 UN: 1477 ADR: 5.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...