Ytterbium(III) bromide hydrate ≥99.9% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals
Warning

14575-09 14575-18
CAAA14575-09EA 175.38 CAD
CAAA14575-09 CAAA14575-18
Ytterbium(III) bromide hydrate ≥99.9% (REO, rare earth oxide basis)
Ytterbium (III) bromide hydrate

MDL: MFCD00049613


Purity: 99.9% CAS No.: 15163-03-8 Molecular Formula: YbBr3.xH2O Formula Weight: 412.77(anhy) Physical Form: Crystalline Sensitivity: Hygroscopic Appearance: White Melting Point: 300° MDL No.: MFCD00049613

Formula: Br₃Yb·H₂O
MW: 430.77
Melting Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00049613
CAS Number: 15163-03-8
EINECS: 237-350-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR