Cesium sulfate ≥99.99% (metals basis)
Supplier: Thermo Scientific Chemicals
Warning
|
14559-14
14559-22
CAAA14559-14EA
137.14
CAD
CAAA14559-14
CAAA14559-22
Cesium sulfate ≥99.99% (metals basis)
Cesium sulphate
MDL: MFCD00010959
Purity: 99.99%
CAS No.: 10294-54-9
Molecular Formula: Cs2SO4
Formula Weight: 361.87
Physical Form: -60 Mesh Powder
Density: 4.243
Melting Point: 1010°
Merck Reference: 14,2016
MDL No.: MFCD00010959
Formula:
Cs₂O₄S Boiling Pt: 1627 °C (1013 hPa) Melting Pt: 1010 °C Density: 4.243 Storage Temperature: Ambient |
MDL Number:
MFCD00010959 CAS Number: 10294-54-9 EINECS: 233-662-6 Merck Index: 14,2016 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...