Aluminium oxide, γ-α-phase ≥99.99% (metals basis)

Supplier: Thermo Scientific Chemicals

14558-14 14558-22 14558-36
CAAA14558-14EA 97.64 CAD
CAAA14558-14 CAAA14558-22 CAAA14558-36
Aluminium oxide, γ-α-phase ≥99.99% (metals basis)
Aluminium oxide, γ-α-phase

MDL: MFCD00003424 Notes: Predominantly ? phase, 10-20% ? phase


Purity: 99.99% CAS No.: 1344-28-1 Molecular Formula: Al2O3 Formula Weight: 101.96 Physical Form: 0.3 Micron Powder, S.A. 15m2/g Odor: Odorless Merck Reference: 14,356 MDL No.: MFCD00003424

Formula: Al₂O₃
Boiling Pt: 2980 °C (1013 hPa)
Melting Pt: 2045 °C
Density: 3.97 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6
Merck Index: 14,356

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR