Aluminium chloride, ultra dry ≥99.99% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Aluminum chloride

14552-06 14552-14 14552-22
CAAA14552-06EA 125.14 CAD
CAAA14552-06 CAAA14552-14 CAAA14552-22
Aluminium chloride, ultra dry ≥99.99% (metals basis)
Aluminium chloride

MDL: MFCD00003422 Fieser: 1,24 12,26 13,15 14,21 15,10 16,10 17,15 18,18 19,11 20,13


Purity: 99.99% CAS No.: 7446-70-0 Molecular Formula: AlCl3 Formula Weight: 133.34 Physical Form: Powder, Ampouled under argon Density: 2.44 pH: 2,4 Melting Point: 190° subl. Merck Reference: 14,337 UN No.: 1726 MDL No.: MFCD00003422

Formula: AlCl₃
MW: 133.34 g/mol
Boiling Pt: 180 °C (1013 hPa)
Melting Pt: 190 °C (subl.)
Density: 2.44
Storage Temperature: Ambient
MDL Number: MFCD00003422
CAS Number: 7446-70-0
EINECS: 231-208-1
UN: 1726
ADR: 8,II
Merck Index: 14,337

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR