Lithium chloride, ultra dry ≥99.9% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
14540-09
14540-18
14540-30
CAAA14540-09EA
123.56
CAD
CAAA14540-09
CAAA14540-18
CAAA14540-30
Lithium chloride, ultra dry ≥99.9% (metals basis)
Lithium chloride
MDL: MFCD00011078
Fieser: 1,609 2,246 4,298 5,677 13,332 16,194 18,211 20,215 21,248
Purity: 99.9%
CAS No.: 7447-41-8
Molecular Formula: LiCl
Formula Weight: 42.39
Physical Form: -10 Mesh Beads, Ampouled under argon
Density: 2.068
Melting Point: 605°
Boiling Point: 1325-1360°
Merck Reference: 14,5528
MDL No.: MFCD00011078
Formula:
LiCl MW: 42.39 g/mol Boiling Pt: 1325…1360 °C Melting Pt: 605 °C Density: 2.068 Storage Temperature: Ambient |
MDL Number:
MFCD00011078 CAS Number: 7447-41-8 EINECS: 231-212-3 Merck Index: 14,5528 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...