Ammonium sulfamate ≥98% ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
In electroplating solutions, making nitrous oxide gas, for manufacture of flameproofing textiles and paper products
Purity: 98% min
CAS No.: 7773-06-0
Molecular Formula: H6N2O3S
Formula Weight: 114.12
Physical Form: Crystalline
Sensitivity: Hygroscopic
Appearance: White
Odor: Weak
pH: 5.2
Melting Point: 131-135°
Boiling Point: 160° dec.
Solubility: Very soluble in water, liquid ammonia. Slightly soluble in ethanol, glycerol, glycol
Merck Reference: 14,554
MDL No.: MFCD00011429
In electroplating solutions, making nitrous oxide gas, for manufacture of flameproofing textiles and paper products
Formula:
H₆N₂O₃S Boiling Pt: 160 °C (dec.) Melting Pt: 131…135 °C Density: >1 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011429 CAS Number: 7773-06-0 EINECS: 231-871-7 Merck Index: 14,554 |
Specification Test Results
Insoluble matter | 0.02% |
Residue after ignition | 0.1% |
Heavy Metals (as Pb) | 5 ppm |
Melting point | 133.0° (2deg) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...