Bismuth(III) telluride ≥99.999% (metals basis) for vacuum deposition
Supplier: Thermo Scientific Chemicals
|
Warning
|
14519-09
14519-18
CAAA14519-09EA
192.76
CAD
CAAA14519-09
CAAA14519-18
Bismuth(III) telluride ≥99.999% (metals basis) for vacuum deposition
Bismuth (III) telluride
MDL: MFCD00014201
Purity: 99.999%
CAS No.: 1304-82-1
Molecular Formula: Bi2Te3
Formula Weight: 800.76
Physical Form: 6mm & Down Pieces
Sensitivity: Moisture Sensitive
Appearance: Grey
Odor: Odorless
Melting Point: 573°
UN No.: 3284
MDL No.: MFCD00014201
Formula:
Bi₂Te₃ Melting Pt: 573 °C Density: 7.69 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00014201 CAS Number: 1304-82-1 EINECS: 215-135-2 UN: 3284 ADR: 6.1,III Merck Index: 12,01331 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...