Titanium nitride ≥99.7% (metals basis)

Supplier: Thermo Scientific Chemicals

Synonyms: Titanium nitride

14510-18 14510-30 14510-A1
CAAA14510-18EA 141.25 CAD
CAAA14510-18 CAAA14510-30 CAAA14510-A1
Titanium nitride ≥99.7% (metals basis)
Titanium nitride

MDL: MFCD00049596


Purity: 99.7% CAS No.: 25583-20-4 Molecular Formula: TiN Formula Weight: 61.91 Physical Form: 10 Micron Powder Density: 5.22 Appearance: Yellow Melting Point: 2950° MDL No.: MFCD00049596

Formula: NTi
Melting Pt: 2950 °C
Density: 5.22
Storage Temperature: Ambient
MDL Number: MFCD00049596
CAS Number: 25583-20-4
EINECS: 247-117-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR