Iron(II) sulfate heptahydrate 99+% ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Iron vitriol, Ferrous sulphate heptahydrate, Ferrous sulfate heptahydrate
14498-30
14498-A1
CAAA14498-30EA
76.47
CAD
CAAA14498-30
CAAA14498-A1
Iron(II) sulfate heptahydrate 99+% ACS
Iron (II) sulphate heptahydrate
MDL: MFCD00149719
Purity: 99+%
CAS No.: 7782-63-0
Molecular Formula: FeO4S.7H2O
Formula Weight: 278.02 (151.91anhy)
Physical Form: Crystalline
Density: 1.897
Appearance: Pale green
Melting Point: 64° -6H2O
Merck Reference: 14,4057
MDL No.: MFCD00149719
Formula:
FeSO₄·7H₂O Melting Pt: 64 °C (-6H₂O) Density: 1.897 Storage Temperature: Ambient |
MDL Number:
MFCD00149719 CAS Number: 7782-63-0 EINECS: 231-753-5 Merck Index: 14,4057 |
Specification Test Results
Insoluble matter | 0.01% |
Cl | 0.001% |
PO3 | 0.001% |
Ca | 0.005% |
Cu | 0.005% |
MG | 0.002% |
K | 0.002% |
NA | 0.0002 |
Ferric iron | 0.001 |
Mn | 0.0005 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...