Ammonium hydrogen sulfate ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

14485-14 14485-22 14485-36
CAAA14485-14EA 85.95 CAD
CAAA14485-14 CAAA14485-22 CAAA14485-36
Ammonium hydrogen sulfate ≥99.9% (metals basis)
Ammonium hydrogen sulphate

MDL: MFCD00051073


Purity: 99.9% CAS No.: 7803-63-6 Molecular Formula: NH4HSO4 Formula Weight: 115.10 Physical Form: Lump Sensitivity: Hygroscopic UN No.: 2506 MDL No.: MFCD00051073

Formula: H₅NO₄S
Boiling Pt: 350 °C (1013 hPa)
Melting Pt: 146.9 °C
Density: 1.787 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00051073
CAS Number: 7803-63-6
EINECS: 232-265-5
UN: 2506
ADR: 8,II
Merck Index: 12,00526

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR