Lithium fluoride, ultra dry ≥99.99% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

14463-09 14463-18
CAAA14463-09EA 252.8 CAD
CAAA14463-09 CAAA14463-18
Lithium fluoride, ultra dry ≥99.99% (metals basis)
Lithium fluoride

MDL: MFCD00011090


Purity: 99.99% CAS No.: 7789-24-4 Molecular Formula: FLi Formula Weight: 25.94 Physical Form: Powder, Ampouled under argon Density: 2.635 Refractive Index: 1.3915 Sensitivity: Hygroscopic Appearance: White Odor: Odorless Melting Point: 845° Boiling Point: 1680° Merck Reference: 14,5531 UN No.: 3288 MDL No.: MFCD00011090

Formula: FLi
MW: 25.94 g/mol
Boiling Pt: 1680 °C
Melting Pt: 845 °C
Density: 2.635
Storage Temperature: Ambient
MDL Number: MFCD00011090
CAS Number: 7789-24-4
EINECS: 232-152-0
UN: 3288
ADR: 6.1 III
Merck Index: 14,5531

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR