Barium carbonate ≥99.95% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
14452-09
14452-18
14452-30
14452-A1
CAAA14452-09EA
70.31
CAD
CAAA14452-09
CAAA14452-18
CAAA14452-30
CAAA14452-A1
Barium carbonate ≥99.95% (metals basis)
Barium carbonate
MDL: MFCD00003448
Notes: m.p. 174°C (90atm), 811°C (1 atm)
Purity: 99.95%
CAS No.: 513-77-9
Molecular Formula: BaCO3
Formula Weight: 197.35
Physical Form: Lump/Powder
Density: 4.43
Melting Point: 811°
Boiling Point: dec.
Merck Reference: 14,969
MDL No.: MFCD00003448
Formula:
BaCO₃ Boiling Pt: Dec. Melting Pt: 811 °C Density: 4.43 Storage Temperature: Ambient |
MDL Number:
MFCD00003448 CAS Number: 513-77-9 EINECS: 208-167-3 UN: 1564 ADR: 6.1,III Merck Index: 14,969 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...