Aluminium ≥99.999% (metals basis), wire, Puratronic®, annealed, Ø 0.25 mm (0.01 in)

Supplier: Thermo Scientific Chemicals

14445-G9 14445-H4
CAAA14445-G9EA 263.86 CAD
CAAA14445-G9 CAAA14445-H4
Aluminium ≥99.999% (metals basis), wire, Puratronic®, annealed, Ø 0.25 mm (0.01 in)
Aluminium

MDL: MFCD00134029


Purity: 99.999% CAS No.: 7429-90-5 Physical Form: ≈0.13g/m, Temper: soft Odor: Odorless MDL No.: MFCD00134029

Formula: Al
Boiling Pt: 2327 °C (1013 hPa)
Melting Pt: 660 °C
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134029
CAS Number: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR