Bismuth ≥99.998% (metals basis), polycrystalline lumps, Puratronic® 2-8 mm (0.08-0.3 in)

Supplier: Thermo Scientific Chemicals

14442-22 14442-36 14442-A3
CAAA14442-22EA 107.76 CAD
CAAA14442-22 CAAA14442-36 CAAA14442-A3
Bismuth ≥99.998% (metals basis), polycrystalline lumps, Puratronic® 2-8 mm (0.08-0.3 in)
Bismuth

Purity: 99.998% CAS No.: 7440-69-9 Odor: Odorless MDL No.: MFCD00134033

Formula: Bi
Boiling Pt: 1490 °C (1013 hPa)
Melting Pt: 271…272 °C
Density: 9.78 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134033
CAS Number: 7440-69-9
EINECS: 231-177-4
Merck Index: 12,01297

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR