Aluminium ≥99.999% (metals basis), wire, Puratronic®, annealed, Ø 2.0 mm (0.08 in)
Supplier: Thermo Scientific Chemicals
14441-G1
14441-G5
14441-G9
CAAA14441-G1EA
85.79
CAD
CAAA14441-G1
CAAA14441-G5
CAAA14441-G9
Aluminium ≥99.999% (metals basis), wire, Puratronic®, annealed, Ø 2.0 mm (0.08 in)
Aluminium
MDL: MFCD00134029
Purity: 99.999%
CAS No.: 7429-90-5
Physical Form: ≈8.4g/m, Temper: soft
Odor: Odorless
MDL No.: MFCD00134029
Formula:
Al Boiling Pt: 2327 °C (1013 hPa) Melting Pt: 660 °C Density: 2.7 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00134029 CAS Number: 7429-90-5 EINECS: 231-072-3 Merck Index: 13,00321 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...