Ytterbium, Plasma Standard Solution, Specpure®, Yb 10000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
14439-AE
14439-AP
CAAA14439-ADEA
158.57
CAD
CAAA14439-AD
CAAA14439-AE
CAAA14439-AP
Ytterbium, Plasma Standard Solution, Specpure®, Yb 10000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Ytterbium Standards
MDL: MFCD00011286
Molecular Formula: Yb2O3 in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011286
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...