Terbium, Plasma Standard Solution, Specpure®, Tb 10000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
14383-AD
14383-AE
14383-AP
CAAA14383-ADEA
110.44
CAD
CAAA14383-AD
CAAA14383-AE
CAAA14383-AP
Terbium, Plasma Standard Solution, Specpure®, Tb 10000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Terbium Standards
MDL: MFCD00011256
Molecular Formula: Tb4O7 in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011256
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...