Cadmium(II) telluride ≥99.999% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Cadmium telluride
14367-06
14367-14
CAAA14367-03EA
57.29
CAD
CAAA14367-03
CAAA14367-06
CAAA14367-14
Cadmium(II) telluride ≥99.999% (metals basis)
Cadmium (II) telluride
Semiconductors, phosphors
Purity: 99.999%
CAS No.: 1306-25-8
Molecular Formula: CdTe
Formula Weight: 240.00
Physical Form: -325 Mesh Polycrystalline Powder
Density: 6.20
Appearance: Black
Odor: Odorless
Melting Point: 1041°
Solubility: Insoluble in water and acids except HNO3 in which it is soluble with decomposition
Merck Reference: 14,1629
UN No.: 3077
MDL No.: MFCD00015998
Semiconductors, phosphors
Formula:
CdTe Melting Pt: 1041 °C Density: 6.20 Storage Temperature: Ambient |
MDL Number:
MFCD00015998 CAS Number: 1306-25-8 EINECS: 215-149-9 UN: 3077 ADR: 9,III Merck Index: 14,1629 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...