Cadmium(II) telluride ≥99.999% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Cadmium telluride
14365-14
14365-22
CAAA14365-06EA
116.34
CAD
CAAA14365-06
CAAA14365-14
CAAA14365-22
Cadmium(II) telluride ≥99.999% (metals basis)
Cadmium (II) telluride
MDL: MFCD00015998
Purity: 99.999%
CAS No.: 1306-25-8
Molecular Formula: CdTe
Formula Weight: 240.00
Physical Form: Polycrystalline Lump
Density: 6.20
Appearance: Black
Odor: Odorless
Melting Point: 1041°
Merck Reference: 14,1629
UN No.: 3077
MDL No.: MFCD00015998
Formula:
CdTe MW: 240.01 g/mol Melting Pt: 1089.85 Density: 6.20 Storage Temperature: Ambient |
MDL Number:
MFCD00015998 CAS Number: 1306-25-8 EINECS: 215-149-9 UN: 3077 ADR: 9 III Merck Index: 14,1629 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...