Niobium ≥99.8% (metals basis), foil, annealed, Thickness: 0.5 mm (0.02 in)

Supplier: Thermo Scientific Chemicals

14358-FY 14358-HL 14358-HP
CAAA14358-FYEA 164.32 CAD
CAAA14358-FY CAAA14358-HL CAAA14358-HP
Niobium ≥99.8% (metals basis), foil, annealed, Thickness: 0.5 mm (0.02 in)
Niobium

MDL: MFCD00011126


Purity: 99.8% CAS No.: 7440-03-1 Physical Form: ≈21.4g/50x100mm Odor: Odorless MDL No.: MFCD00011126

Formula: Nb
Boiling Pt: 3700 °C (1013 hPa)
Melting Pt: 2468 °C
Density: 8.57 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011126
CAS Number: 7440-03-1
EINECS: 231-113-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR