Strontium carbonate 99% (max. 1% Ba)

Supplier: Thermo Scientific Chemicals

14343-30 14343-A1
CAAA14343-30EA 113.29 CAD
CAAA14343-30 CAAA14343-A1
Strontium carbonate 99% (max. 1% Ba)
Strontium carbonate

MDL: MFCD00011250 Notes: 1340°C -CO2


Purity: 99% CAS No.: 1633-05-2 Molecular Formula: SrCO3 Formula Weight: 147.63 Physical Form: -325 Mesh, Typically 1 Micron Powder Density: 3.70 Melting Point: 1100° dec. Merck Reference: 14,8838 MDL No.: MFCD00011250

Formula: CO₃Sr
MW: 147.63 g/mol
Boiling Pt: 2647 °C (1 mmHg)
Melting Pt: 1100 °C (dec.)
Density: 3.70
Storage Temperature: Ambient
MDL Number: MFCD00011250
CAS Number: 1633-05-2
EINECS: 216-643-7
Merck Index: 14,8838

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR