Barium carbonate ≥99.8%
Supplier: Thermo Scientific Chemicals
|
Warning
|
14341-18
14341-30
14341-A1
14341-A7
CAAA14341-18EA
61.3
CAD
CAAA14341-18
CAAA14341-30
CAAA14341-A1
CAAA14341-A7
Barium carbonate ≥99.8%
Barium carbonate
MDL: MFCD00003448
Notes: m.p. 174°C (90 atm), 811°C (1 atm)
Purity: 99.8%
CAS No.: 513-77-9
Molecular Formula: BaCO3
Formula Weight: 197.35
Physical Form: 1 Micron Powder
Density: 4.43
Melting Point: 811°
Boiling Point: dec.
Merck Reference: 14,969
MDL No.: MFCD00003448
Formula:
BaCO₃ MW: 197.34 g/mol Boiling Pt: Dec. Melting Pt: 1350 Density: 4.43 Storage Temperature: Ambient |
MDL Number:
MFCD00003448 CAS Number: 513-77-9 EINECS: 208-167-3 UN: 1564 ADR: 6.1 III Merck Index: 14,969 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...