Manganese(IV) oxide Mn ≥ 58%, tech., activated
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Manganese dioxide, Manganese(IV) oxide, activated, Manganese peroxide, Mangan(IV)-oxid
As an oxidizing agent, source of metallic manganese, decolorizing glass, painting porcelain, as an analytical reagent
CAS No.: 1313-13-9
Molecular Formula: MnO2
Formula Weight: 86.94
Physical Form: Powder
Density: 5.026
Appearance: Black
Melting Point: 535° dec.
Solubility: Insoluble in water, nitric acid, sulfuric acid. Soluble in HCl. In presence of hydrogen peroxide or oxalic acid soluble in dilute HNO3 and H2SO4
Merck Reference: 14,5730
MDL No.: MFCD00003463
As an oxidizing agent, source of metallic manganese, decolorizing glass, painting porcelain, as an analytical reagent
Formula:
MnO₂ MW: 86.94 g/mol Melting Pt: 535 (dec.) Density: 5.026 Storage Temperature: Ambient |
MDL Number:
MFCD00003463 CAS Number: 1313-13-9 EINECS: 215-202-6 Merck Index: 14,05730 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...