Niobium ≥99.96% (metals basis excluding Ta), wire, Ø 0.5 mm (0.02 in)

Supplier: Thermo Scientific Chemicals

14335-BU 14335-BY 14335-CC
CAAA14335-BUEA 83.74 CAD
CAAA14335-BU CAAA14335-BY CAAA14335-CC
Niobium ≥99.96% (metals basis excluding Ta), wire, Ø 0.5 mm (0.02 in)
Niobium

MDL: MFCD00011126


Purity: 99.96% CAS No.: 7440-03-1 Physical Form: ≈1.68g/m Odor: Odorless MDL No.: MFCD00011126

Formula: Nb
Boiling Pt: 3700 °C (1013 hPa)
Melting Pt: 2468 °C
Density: 8.57 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011126
CAS Number: 7440-03-1
EINECS: 231-113-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR