Manganese(II) carbonate Mn ≥ 44%
Supplier: Thermo Scientific Chemicals
14324-36
14324-A3
CAAA14324-36EA
87.06
CAD
CAAA14324-36
CAAA14324-A3
Manganese(II) carbonate Mn ≥ 44%
Manganese (II) carbonate
Pigment, varnish drier
CAS No.: 598-62-9
Molecular Formula: CMnO3
Formula Weight: 114.95
Physical Form: -200 Mesh Powder
Density: 3.125
Appearance: Light brown to violet
Odor: Odorless
Melting Point: 350° dec.
Solubility: Insoluble in water, alcohol. Soluble in dilute inorganic acids
Merck Reference: 14,5726
MDL No.: MFCD00011116
Pigment, varnish drier
Formula:
CMnO₃ Melting Pt: 350 °C (dec.) Density: 3.125 Storage Temperature: Ambient |
MDL Number:
MFCD00011116 CAS Number: 598-62-9 EINECS: 209-942-9 Merck Index: 14,5726 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...