Potassium thiocyanate ≥98.5% ACS

Supplier: Thermo Scientific Chemicals
Warning

14318-22 14318-36
CAAA14318-22EA 88.8 CAD
CAAA14318-22 CAAA14318-36
Potassium thiocyanate ≥98.5% ACS
Potassium thiocyanate

MDL: MFCD00011413 Beilstein Registry No.: 3594799 Soluble in water, acetone, and alcohol. When dissolved in its own weight of water, the temperature drops ≈30°C Fieser: 1,954


Purity: 98.5% min CAS No.: 333-20-0 Molecular Formula: KSCN Formula Weight: 97.18 Physical Form: Crystalline Density: 1.89 pH: 5.3-8.5 Melting Point: 172-175° Boiling Point: 500° dec. Solubility: Soluble in water, acetone, and alcohol. When dissolved in its own weight of water, the temperature drops ≈30° Beilstein Reference: 3594799 Merck Reference: 14,7691 MDL No.: MFCD00011413

Formula: KSCN
Boiling Pt: 500 °C (dec.)
Melting Pt: 172…175 °C
Density: 1.89
Storage Temperature: Ambient
MDL Number: MFCD00011413
CAS Number: 333-20-0
EINECS: 206-370-1
Merck Index: 14,7691

Order Now

Specification Test Results

Insoluble in water 0.005%
pH of a 5 % solution 5.3-8.7 at 25°
Cl 0.005%
SO3 0.005%
NH3 0.003%
Heavy Metals (as Pb) 5 ppm
Fe 2 ppm
NA 0.005%
Iodine-consuming substances Pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR