Potassium sulfate ≥99.0% ACS
Supplier: Thermo Scientific Chemicals
14311-36
14311-A3
CAAA14311-36EA
112.65
CAD
CAAA14311-36
CAAA14311-A3
Potassium sulfate ≥99.0% ACS
Potassium sulphate
MDL: MFCD00011388
Soluble in water. Insoluble in alcohol.
Purity: 99.0% min
CAS No.: 7778-80-5
Molecular Formula: K2SO4
Formula Weight: 174.27
Physical Form: Crystalline
Density: 2.662
Melting Point: 1069°
Boiling Point: 1689°
Solubility: Soluble in water. Insoluble in alcohol
Merck Reference: 14,7674
MDL No.: MFCD00011388
Formula:
K₂SO₄ Boiling Pt: 1689 °C Melting Pt: 1069 °C Density: 2.662 Storage Temperature: Ambient |
MDL Number:
MFCD00011388 CAS Number: 7778-80-5 EINECS: 231-915-5 Merck Index: 14,7674 |
Specification Test Results
Insoluble matter | 0.01% |
pH of a 5 % solution | 5.5-8.5 at 25° |
Cl | 0.001% |
Nitrogen compounds (as N) | 5 ppm |
Ca | 0.01% |
MG | 0.005% |
Heavy Metals (as Pb) | 5 ppm max. |
Fe | 5 ppm |
NA | 0.02%% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...