Calcium acetylide

Supplier: Thermo Scientific Chemicals
Danger

14308-A1 14308-D9
CAAA14308-A1EA 103.33 CAD
CAAA14308-A1 CAAA14308-D9
Calcium acetylide
Calcium acetylide

MDL: MFCD00010905 Beilstein Registry No.: 3909011


CAS No.: 75-20-7 Molecular Formula: C2Ca Formula Weight: 64.09 Physical Form: 9-40mm Lump Density: 2.22 Sensitivity: Moisture Sensitive Appearance: Greyish black Melting Point: 2300° Beilstein Reference: 3909011 Merck Reference: 14,1656 UN No.: 1402 MDL No.: MFCD00010905

Formula: CaC₂
Melting Pt: 2300 °C
Density: 2.22
Storage Temperature: Ambient
MDL Number: MFCD00010905
CAS Number: 75-20-7
EINECS: 200-848-3
UN: 1402
ADR: 4.3,II
Merck Index: 14,1656

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR