Copper(II) sulfide, ultra dry ≥99.98% (metals basis)

Supplier: Thermo Scientific Chemicals

14300-09 14300-18
CAAA14300-09EA 461.36 CAD
CAAA14300-09 CAAA14300-18
Copper(II) sulfide, ultra dry ≥99.98% (metals basis)
Copper (II) sulphide

MDL: MFCD00016066


Purity: 99.98% CAS No.: 1317-40-4 Molecular Formula: CuS Formula Weight: 95.60 Physical Form: Powder, Ampouled under argon Density: 3.9-4.6 Refractive Index: 1.45 Sensitivity: Air and Moisture Sensitive Odor: Odorless Melting Point: 220° dec. Merck Reference: 14,2965 MDL No.: MFCD00016066

Formula: CuS
Melting Pt: 220 °C (dec.)
Density: 4.6
Storage Temperature: Ambient
MDL Number: MFCD00016066
CAS Number: 1317-40-4
EINECS: 215-271-2
Merck Index: 14,2965

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR