Cesium bromide, ultra dry ≥99.9% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Cesiumbromide
14289-09
14289-18
CAAA14289-09EA
267.02
CAD
CAAA14289-09
CAAA14289-18
Cesium bromide, ultra dry ≥99.9% (metals basis)
Cesium bromide
MDL: MFCD00010954
Purity: 99.9%
CAS No.: 7787-69-1
Molecular Formula: CsBr
Formula Weight: 212.81
Physical Form: -10 Mesh Beads, Ampouled under argon
Density: 4.44
Refractive Index: 1.6984
Sensitivity: Hygroscopic
Appearance: White
Odor: Odorless
Melting Point: 636°
Boiling Point: 1300°
Merck Reference: 14,2009
MDL No.: MFCD00010954
Formula:
BrCs MW: 212.81 g/mol Boiling Pt: 1300 °C Melting Pt: 636 °C Density: 4.44 Storage Temperature: Ambient |
MDL Number:
MFCD00010954 CAS Number: 7787-69-1 EINECS: 232-130-0 Merck Index: 14,2009 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...