Magnesium iodide, ultra dry ≥99.996% (metals basis)
Supplier: Thermo Scientific Chemicals
14283-03
14283-09
CAAA14283-03EA
124.66
CAD
CAAA14283-03
CAAA14283-09
Magnesium iodide, ultra dry ≥99.996% (metals basis)
Magnesium iodide
MDL: MFCD00049485
Purity: 99.996%
CAS No.: 10377-58-9
Molecular Formula: I2Mg
Formula Weight: 278.12
Physical Form: -10 Mesh Beads, Ampouled under argon
Sensitivity: Light Sensitive and Hygroscopic
Appearance: White
Odor: Odorless
Melting Point: 637° dec.
Merck Reference: 14,5671
MDL No.: MFCD00049485
Formula:
MgI₂ Melting Pt: 637 °C (dec.) Density: 3.89 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00049485 CAS Number: 10377-58-9 EINECS: 233-825-1 Merck Index: 14,5671 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...