Neodymium trichloride, ultra dry ≥99.99% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Neodymium (III) chloride

14282-06 14282-14
CAAA14282-06EA 383.94 CAD
CAAA14282-06 CAAA14282-14
Neodymium trichloride, ultra dry ≥99.99% (REO, rare earth oxide basis)
Neodymium trichloride

MDL: MFCD00011132


Purity: 99.99% CAS No.: 10024-93-8 Molecular Formula: NdCl3 Formula Weight: 250.60 Physical Form: -20 Mesh Powder, Ampouled under argon Density: 4.134 Melting Point: 784° Merck Reference: 14,6450 MDL No.: MFCD00011132

Formula: Cl₃Nd
MW: 250.6 g/mol
Boiling Pt: 1600
Melting Pt: 758
Density: 4.134
Storage Temperature: Ambient
MDL Number: MFCD00011132
CAS Number: 10024-93-8
EINECS: 233-031-5
Merck Index: 14,6450

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR