Sodium chlorite tech. ∼80%, Technical Grade

Supplier: Thermo Scientific Chemicals
Danger

14265-36 14265-A3 14265-A9
CAAA14265-36EA 100.49 CAD
CAAA14265-36 CAAA14265-A3 CAAA14265-A9
Sodium chlorite tech. ∼80%, Technical Grade
Sodium chlorite

MDL: MFCD00003478


Purity: 80% CAS No.: 7758-19-2 Molecular Formula: ClNaO2 Formula Weight: 90.44 Physical Form: Flake/Powder Sensitivity: Hygroscopic Appearance: White Odor: Odorless Melting Point: 180-200° dec. Merck Reference: 14,8600 UN No.: 1496 MDL No.: MFCD00003478

Formula: ClNaO₂
MW: 90.44 g/mol
Boiling Pt: 112 °C (1013 hPa)
Melting Pt: 180…200 °C (dec.)
Density: 1.28 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003478
CAS Number: 7758-19-2
EINECS: 231-836-6
UN: 1496
ADR: 5.1,II
Merck Index: 14,8600

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR