Potassium ethanolate ≥95%

Supplier: Thermo Scientific Chemicals
Danger

14263-30 14263-A1
CAAA14263-30EA 214.88 CAD
CAAA14263-30 CAAA14263-A1
Potassium ethanolate ≥95%
Potassium ethanolate

MDL: MFCD00054277 Soluble in ethanol, ether


Purity: 95% CAS No.: 917-58-8 Molecular Formula: C2H5KO Formula Weight: 84.16 Physical Form: Powder Sensitivity: Air and Moisture Sensitive Appearance: Pale yellow to pale brown Odor: Alcohol-like Melting Point: 250° dec. Solubility: Soluble in ethanol, ether UN No.: 3206 MDL No.: MFCD00054277

Formula: C₂H₅KO
Melting Pt: 250 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00054277
CAS Number: 917-58-8
EINECS: 213-029-0
UN: 3206
ADR: 4.2,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR