Potassium methanolate 90+%

Supplier: Thermo Scientific Chemicals
Danger

14261-18 14261-30
CAAA14261-18EA 69.05 CAD
CAAA14261-18 CAAA14261-30
Potassium methanolate 90+%
Potassium methanolate

MDL: MFCD00012416


Purity: 90+% CAS No.: 865-33-8 Molecular Formula: CH3KO Formula Weight: 70.14 Physical Form: Powder Sensitivity: Air and Moisture Sensitive UN No.: 3206 MDL No.: MFCD00012416

Formula: CH₃KO
Density: 0,85 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012416
CAS Number: 865-33-8
EINECS: 212-736-1
UN: 3206
ADR: 4.2,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR