Silver ≥99.9% (metals basis), tubing 6.35 mm (0.25 in) OD, 4.35 mm (0.171 in) ID

Supplier: Thermo Scientific Chemicals

14259-BS 14259-BW 14259-VC
CAAA14259-BSEA 178.54 CAD
CAAA14259-BS CAAA14259-BW CAAA14259-VC
Silver ≥99.9% (metals basis), tubing 6.35 mm (0.25 in) OD, 4.35 mm (0.171 in) ID
Silver

MDL: MFCD00003397


Purity: 99.9% CAS No.: 7440-22-4 Physical Form: ≈18g/10cm, Temper: as drawn Odor: Characteristic MDL No.: MFCD00003397

Formula: Ag
MW: 107.87 g/mol
Boiling Pt: 2212
Melting Pt: 962
Density: 10.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR