Copper(II) bromide Cu ≥ 28,1%, REacton®, Reagent Grade

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Cupric bromide, Cuprous bromide

14258-22 14258-36
CAAA14258-22EA 97.96 CAD
CAAA14258-22 CAAA14258-36
Copper(II) bromide Cu ≥ 28,1%, REacton®, Reagent Grade
Copper (II) bromide

MDL: MFCD00010970 Fieser: 1,161 2,84 5,158 6,138 14,100 15,100 18,111 19,106 21,143


CAS No.: 7789-45-9 Molecular Formula: CuBr2 Formula Weight: 223.36 Physical Form: Crystalline Density: 4.710 Sensitivity: Hygroscopic Appearance: Dark grey to black Odor: Odorless Melting Point: 498° Boiling Point: 900° Merck Reference: 14,2629 UN No.: 3260 MDL No.: MFCD00010970

Formula: Br₂Cu
MW: 223.35 g/mol
Boiling Pt: 900
Melting Pt: 498
Density: 4.710
Storage Temperature: Ambient
MDL Number: MFCD00010970
CAS Number: 7789-45-9
EINECS: 232-167-2
UN: 3260
ADR: 8 III
Merck Index: 14,2629

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR