Silver ≥99.9% (metals basis), tubing 6.35 mm (0.25 in) OD, 5.84 mm (0.230 in) ID

Supplier: Thermo Scientific Chemicals

14252-BS 14252-BW 14252-VC
CAAA14252-BSEA 119.29 CAD
CAAA14252-BS CAAA14252-BW CAAA14252-VC
Silver ≥99.9% (metals basis), tubing 6.35 mm (0.25 in) OD, 5.84 mm (0.230 in) ID
Silver

MDL: MFCD00003397


Purity: 99.9% CAS No.: 7440-22-4 Physical Form: ≈5.1g/10cm, Temper: as drawn Odor: Characteristic MDL No.: MFCD00003397

Formula: Ag
Boiling Pt: 2212 °C (1013 hPa)
Melting Pt: 962 °C
Density: 10.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR