Ammonium hydrogen carbonate ≥99.0%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Ammonium bicarbonate
In cooling baths, fire extinguishing compounds, baking powder formulations, and in the manufacture of ceramics and pigments. Also used for compost treatment to accelerate decomposition
Purity: 99.0%
CAS No.: 1066-33-7
Molecular Formula: CH5NO3
Formula Weight: 79.06
Physical Form: Powder
Density: 1.586
Appearance: White
Odor: Like ammonia
Melting Point: ca 60° dec.
Solubility: Soluble in cold water but decomposed by hot water. Insoluble in alcohol and acetone
Beilstein Reference: 4329606
Merck Reference: 14,497
MDL No.: MFCD00012138
In cooling baths, fire extinguishing compounds, baking powder formulations, and in the manufacture of ceramics and pigments. Also used for compost treatment to accelerate decomposition
Formula:
NH₄HCO₃ MW: 79.06 g/mol Melting Pt: ∼ 60 °C (dec.) Density: 1.586 Storage Temperature: Ambient |
MDL Number:
MFCD00012138 CAS Number: 1066-33-7 EINECS: 213-911-5 Merck Index: 14,497 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...