Lead(II) nitrate ≥99.0% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Lead dinitrate
As an oxidizer in dye industry, in manufacturing of mother-of-pearl, as mordant in printing textile, as sensitizer in photography
Purity: 99.0% min
CAS No.: 10099-74-8
Molecular Formula: Pb(NO3)2
Formula Weight: 331.20
Physical Form: Crystalline
Density: 4.530
Appearance: White
Odor: Odorless
Melting Point: 470° dec.
Solubility: Freely soluble in water. Soluble in absolute alcohol, methanol. Insoluble in concentrated HNO3
Merck Reference: 14,5414
UN No.: 1469
MDL No.: MFCD00011153
As an oxidizer in dye industry, in manufacturing of mother-of-pearl, as mordant in printing textile, as sensitizer in photography
Formula:
Pb(NO₃)₂ MW: 331.21 g/mol Melting Pt: 470 °C (dec.) Density: 4.530 Storage Temperature: Ambient |
MDL Number:
MFCD00011153 CAS Number: 10099-74-8 EINECS: 233-245-9 UN: 1469 ADR: 5.1 II Merck Index: 14,5414 |
Specification Test Results
Form | Crystalline |
Cl | 0.1% |
Ca | 0.005% |
Cu | 0.002% |
Fe | 0.001% |
K | 0.005% |
NA | 0.02% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...